6-Amino-2-naphthoesäure 90%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
6-Amino-2-naphthoic acid, 6-Amino-2-naphthalenecarboxylic acid, 6-Amino-2-naphthoic Acid
344500050
ACRO344500050EA
160
EUR
ACRO344500050
6-Amino-2-naphthoesäure 90%
6-Amino-2-naphthoesäure
Formel:
C₁₁H₉NO₂ Molecular Weight: 187,1947 g/mol Schmelzpunkt: 222…227 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01861831 CAS-Nummer: 116668-47-4 |
Spezifikation Testergebnisse
Appearance | Green-beige to brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥88 % |
Titration with HClO4 | ≥88 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...