3-Aminopropylphosphonic acid 97%

Lieferant: Thermo Fisher Scientific
Achtung

344940010 344940050
ACRO344940010EA 198 EUR
ACRO344940010 ACRO344940050
3-Aminopropylphosphonic acid 97%
3-Aminopropylphosphonic acid

CAS No.: 13138-33-5

Formel: C3H10NO3P
Molecular Weight: 139.0902 g/mol
Schmelzpunkt: 290…294 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008222
CAS-Nummer: 13138-33-5

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR