2',6'-Dichlor-3'-fluoracetophenon 97%

Lieferant: Thermo Scientific
Achtung

345511000
ACRO345511000EA 331 EUR
ACRO345511000
2',6'-Dichlor-3'-fluoracetophenon 97%
2',6'-Dichlor-3'-fluoracetophenon
Formel: C₈H₅Cl₂FO
Molecular Weight: 207,03 g/mol
Flammpunkt: 110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093760
CAS-Nummer: 290835-85-7

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5275 to 1.5295 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR