Vitamin A acetat 1,5 MIU/g in Sonnenblumenöl stabilisiert

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: alle-trans-Retinylacetat

346321000 346325000
ACRO346321000EA 130 EUR
ACRO346321000 ACRO346325000
Vitamin A acetat 1,5 MIU/g in Sonnenblumenöl stabilisiert
alle-trans-Retinylacetat
Formel: C₂₂H₃₂O₂
Molecular Weight: 328.49 g/mol
Flammpunkt: >100 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00019413
CAS-Nummer: 127-47-9
UN: 3082
ADR: 9 III

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to brown-yellow Partly crystalline when cooled
Assay 1.50 to 1.65 million IU/g (Vitamin A)
Identification (TLC): conforms
Stabilizer BHT
Related substances ≤0.14 (A370/A326)
Acid value ≤2.0 mg KOH/g
Peroxide value ≤15 meq/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR