Ethylene glycol monostearate

Lieferant: Thermo Fisher Scientific

346951000 346952500
ACRO346951000EA 30.3 EUR
ACRO346951000 ACRO346952500
Ethylene glycol monostearate
Ethylene glycol monostearate

Appearance: White to light yellow Flakes

Formel: C₂₀H₄₀O₃
Siedepunkt: 149 °C
Schmelzpunkt: 55…60 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051465
CAS-Nummer: 111-60-4

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Flakes
Infrared spectrum Conforms
Melting point 55 to 60 °C
Additional info compound is a mixture of various chain-length esters and di(ethylene glycol) ester


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR