8-Hydroxy-2-chinolincarbaldehyd 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 8-Hydroxyquinoline-2-carboxaldehyde

347630050 347630010
ACRO347630050EA 319 EUR
ACRO347630050 ACRO347630010
8-Hydroxy-2-chinolincarbaldehyd 98%
8-Hydroxyquinoline-2-carbaldehyde

Appearance: Yellow Crystalline powder

Formel: C₁₀H₇NO₂
Molecular Weight: 173.1681 g/mol
Schmelzpunkt: 96…100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00168962
CAS-Nummer: 14510-06-6

Bestellen

Spezifikation Testergebnisse

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 96 to 100 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR