Furfuryl glycidyl ether 97%

Lieferant: Thermo Fisher Scientific

Synonyme: 2-((Oxiran-2-ylmethoxy)methyl)furan

348490050 348490250
ACRO348490050EA 91.9 EUR
ACRO348490050 ACRO348490250
Furfuryl glycidyl ether 97%
Furfuryl glycidyl ether

Appearance: Clear yellow to brown Liquid

Formel: C₈H₁₀O₃
Molecular Weight: 154.165 g/mol
Siedepunkt: 103…104 °C (11 mmHg)
Dichte: 1.122 g/cm³
Flammpunkt: 102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013348
CAS-Nummer: 5380-87-0

Bestellen

Spezifikation Testergebnisse

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4800 to 1.4820 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR