3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluordecyl-acrylat 97% stabilisiert

Lieferant: Thermo Scientific
Achtung

Synonyme: Acrylic Acid 1H,1H,2H,2H-Heptadecafluorodecyl Ester

348610050
ACRO348610050EA 50.1 EUR
ACRO348610050
3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluordecyl-acrylat 97% stabilisiert
1H,1H,2H,2H-Heptadecafluorodecyl acrylate

Appearance: Clear colorless to light yellow Liquid

Formel: C₁₃H₇F₁₇O₂
Molecular Weight: 518,17 g/mol
Siedepunkt: 100 °C (4 mmHg)
Flammpunkt: >110 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00042306
CAS-Nummer: 27905-45-9

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3360 to 1.3380 (20°C, 589 nm)
GC ≥96.0 %
Stabilizer Approx. 15 ppm TBC and 70 ppm BHT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR