2-(2,6-Dichlorphenyl)ethanol 96%

Lieferant: Thermo Scientific

Synonyme: 2,6-Dichlorophenethyl Alcohol

350760050
ACRO350760050EA 54.5 EUR
ACRO350760050
2-(2,6-Dichlorphenyl)ethanol 96%
2-(2,6-Dichlorophenyl)ethanol

Appearance: White to light yellow Crystalline powder or crystals or flakes

Formel: C₈H₈Cl₂O
Molecular Weight: 191,05 g/mol
Schmelzpunkt: 59…65 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00800674
CAS-Nummer: 30595-79-0

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder or crystals or flakes
Infrared spectrum Conforms
Melting point 74 to 79 °C
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR