2-Methylphenethylamin 97%

Lieferant: Thermo Fisher Scientific
Gefahr

351100050
ACRO351100050EA 130 EUR
ACRO351100050
2-Methylphenethylamin 97%
2-Methylphenethylamin
Formel: C₉H₁₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD01310827
CAS-Nummer: 55755-16-3
UN: 2735
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5340 to 1.5360 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR