3-Methylphenethylamin 98%

Lieferant: Thermo Fisher Scientific
Gefahr

351110010 351110050
ACRO351110010EA 29.2 EUR
ACRO351110010 ACRO351110050
3-Methylphenethylamin 98%
3-Methylphenethylamin
Formel: C₉H₁₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD01310828
CAS-Nummer: 55755-17-4
UN: 2735
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR