2-Naphthalinmethanol 98%

Lieferant: Thermo Fisher Scientific

351150010 351150050 351150250
ACRO351150010EA 36.4 EUR
ACRO351150010 ACRO351150050 ACRO351150250
2-Naphthalinmethanol 98%
2-Naphthalinmethanol
Formel: C₁₁H₁₀O
Molecular Weight: 158.2000 g/mol
Schmelzpunkt: 79…83 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004124
CAS-Nummer: 1592-38-7

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 79 to 83 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR