Benzyl-2,2,2-trichlorethanimidat 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Trichloroacetimidic Acid Benzyl Ester, Benzyl 2,2,2-trichloroacetimidate
351600250
351601000
ACRO351600250EA
93.9
EUR
ACRO351600250
ACRO351601000
Benzyl-2,2,2-trichlorethanimidat 99%
Benzyl-2,2,2-trichlorethanimidat
CAS No.: 81927-55-1
Formel:
C₉H₈Cl₃NO Molecular Weight: 252.52 g/mol Siedepunkt: 109…113 °C (.5 mmHg) Dichte: 1.359 g/cm³ Flammpunkt: >110 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00000805 CAS-Nummer: 81927-55-1 |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5435 to 1.5455 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...