Kaliumtetraoxalat Dihydrat 99%, reinst
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Oxalic acid, potassium salt dihydrate, Potassium trihydrogen dioxalate dihydrate
352111000
352110010
352115000
ACRO352111000EA
27.4
EUR
ACRO352111000
ACRO352110010
ACRO352115000
Kaliumtetraoxalat Dihydrat 99%, reinst
Kaliumtetraoxalat Dihydrat
Formel:
C₄H₃KO₈·2H₂O Molecular Weight: 254.19 g/mol (218.16 anhy) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150443 CAS-Nummer: 6100-20-5 EINECS: 204-874-6 |
Spezifikation Testergebnisse
Appearance | Colorless to white Crystalline powder or crystals |
Infrared spectrum | Conforms |
Loss on drying | 12 % to 16 % (1 g, 105°C) |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...