Bromcyan 3 M in Dichlormethan

Lieferant: Thermo Scientific
Gefahr

Synonyme: Cyanogenbromid

353991000
ACRO353991000EA 103 EUR
ACRO353991000
Bromcyan 3 M in Dichlormethan
Bromcyan
Formel: CBrN
Molecular Weight: 105,92 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00011597
CAS-Nummer: 506-68-3
EINECS: 208-051-2
UN: 2927
ADR: 6.1,I

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Solution
Strength 2.95 to 3.3 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR