Morin Hydrat

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2',3,4',5,7-Pentahydroxyflavone

354000100 354000050 354000500
ACRO354000100EA 93.9 EUR
ACRO354000100 ACRO354000050 ACRO354000500
Morin Hydrat
Morin Hydrat

CAS No.: 654055-01-3

Formel: C₁₅H₁₀O₇
Molecular Weight: 302.24 g/mol
Schmelzpunkt: 300 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00217054
CAS-Nummer: 654055-01-3
UN: 3077
ADR: 9 III

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to brown Powder
Infrared spectrum Conforms
UV spectrum Complies
Water ≤7 %
Sulfated ash ≤2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR