2-(4-Methoxyphenyl)ethanol 96%

Lieferant: Thermo Fisher Scientific

Synonyme: 2-(4-Methoxyphenyl)ethylalkohol, 4-Methoxyphenethylalkohol

354050100 354050500
ACRO354050100EA 73.2 EUR
ACRO354050100 ACRO354050500
2-(4-Methoxyphenyl)ethanol 96%
2-(4-Methoxyphenyl)ethanol
Formel: C₉H₁₂O₂
Schmelzpunkt: 28…30 °C
Flammpunkt: >112 °C
MDL: MFCD00002900
CAS-Nummer: 702-23-8
EINECS: 211-866-6

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light brown Liquid or solid
Infrared spectrum Conforms
Refractive index 1.5360 to 1.5380 (20°C, 589 nm)
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR