4-(Trifluormethyl)thiophenol 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-(Trifluormethyl)phenylmercaptan, 4-Mercaptobenzotrifluorid, 4-(Trifluormethyl)benzolthiol
355290010
ACRO355290010EA
65.8
EUR
ACRO355290010
4-(Trifluormethyl)thiophenol 97%
4-(Trifluormethyl)thiophenol
Formel:
C₇H₅F₃S Molecular Weight: 178,18 g/mol Siedepunkt: 68…72 °C (15 mmHg) Dichte: 1,31 g/cm³ (25 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00042320 CAS-Nummer: 825-83-2 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4920 to 1.4950 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...