Methyl-5-brom-2-furoat 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
355750010
355750050
355750250
ACRO355750010EA
33.7
EUR
ACRO355750010
ACRO355750050
ACRO355750250
Methyl-5-brom-2-furoat 98%
Methyl-5-brom-2-furoat
Formel:
C₆H₅BrO₃ Schmelzpunkt: 65…67 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00092313 CAS-Nummer: 2527-99-3 |
Spezifikation Testergebnisse
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 63 to 68 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...