6-Brompiperonal 98%

Lieferant: Thermo Fisher Scientific
Achtung

355920010 355920050 355920250
ACRO355920010EA 29.7 EUR
ACRO355920010 ACRO355920050 ACRO355920250
6-Brompiperonal 98%
6-Brompiperonal
Formel: C₈H₅BrO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00022952
CAS-Nummer: 15930-53-7

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 125 to 129 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR