(R)-(+)-2,2'-Diamino-1,1'-binaphthalene 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
356510010
ACRO356510010EA
145
EUR
ACRO356510010
(R)-(+)-2,2'-Diamino-1,1'-binaphthalene 99%
(R)-(+)-2,2'-Diamino-1,1'-binaphthalene
CAS No.: 18741-85-0
Formel:
C₂₀H₁₆N₂ Schmelzpunkt: 242…244 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00145204 CAS-Nummer: 18741-85-0 |
Spezifikation Testergebnisse
Appearance | Off-white to brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥98.5 % |
Specific optical rotation | +153° to +159° (20°C, 589 nm) (c=1, pyridine) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...