(S)-(-)-2,2'-Diamino-1,1'-binaphthalene 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: (S)-1,1'-Binaphthyl-2,2'-diamine, (S)-(-)-BINAM, (S)-(-)-1,1'-Binaphthyl-2,2'-diamine, (S)-(-)-1,1'-Bi(2-naphthylamine), (S)-(-)-2,2'-Diamino-1,1'-binaphthyl

356520010
ACRO356520010EA 147 EUR
ACRO356520010
(S)-(-)-2,2'-Diamino-1,1'-binaphthalene 97%
(S)-(-)-2,2'-Diamino-1,1'-binaphthalene

CAS No.: 18531-95-8

Formel: C₂₀H₁₆N₂
Molecular Weight: 284,362 g/mol
Schmelzpunkt: 242…244 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00145204
CAS-Nummer: 18531-95-8

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Specific optical rotation -149.5° to -155.5° (20°C, 589 nm) (c=1, pyridine)
Titration with HClO4 96.0 to 104.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR