Cycloheximid 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Naramycin A, CHX, 4-[(2R)-2-[(1S,3S,5S)-3,5-Dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidin-2,6-dion, Actidion
357420010
357420050
357420250
ACRO357420010EA
87.7
EUR
ACRO357420010
ACRO357420050
ACRO357420250
Cycloheximid 95%
Cycloheximid
Formel:
C₁₅H₂₃NO₄ Schmelzpunkt: 107…114 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082346 CAS-Nummer: 66-81-9 EINECS: 200-636-0 UN: 2811 ADR: 6.1,I |
|
Spezifikation Testergebnisse
Appearance | White to light beige Powder |
Infrared spectrum | Conforms |
Melting point | 108 to 118 °C |
HPLC | ≥94 % |
Specific optical rotation | -25° to -32° (20 °C, 589 nm) (c=1, CHCl3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...