5-Brom-2-hydroxypyridin 99%

Lieferant: Thermo Fisher Scientific
Achtung

357640050 357640250
ACRO357640050EA 46 EUR
ACRO357640050 ACRO357640250
5-Brom-2-hydroxypyridin 99%
5-Brom-2-hydroxypyridin
Formel: C₅H₄BrNO
Schmelzpunkt: 179…182 °C
MDL: MFCD00234042
CAS-Nummer: 13466-38-1

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to yellow-brown Crystalline powder
Infrared spectrum Conforms
Melting point 179 to 182 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR