5-Brom-2-hydroxypyridin 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
357640050
357640250
ACRO357640050EA
46
EUR
ACRO357640050
ACRO357640250
5-Brom-2-hydroxypyridin 99%
5-Brom-2-hydroxypyridin
Formel:
C₅H₄BrNO Schmelzpunkt: 179…182 °C |
MDL:
MFCD00234042 CAS-Nummer: 13466-38-1 |
Spezifikation Testergebnisse
Appearance | Off-white to yellow-brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 179 to 182 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...