4-Iod-m-toluidin 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
357800050
357800250
357801000
ACRO357800050EA
35.4
EUR
ACRO357800050
ACRO357800250
ACRO357801000
4-Iod-m-toluidin 98%
4-Iod-m-toluidin
Formel:
C₇H₈IN Schmelzpunkt: 50…55 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01569451 CAS-Nummer: 4949-69-3 |
Spezifikation Testergebnisse
Appearance | Beige to light brown to dark grey Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 50 to 55 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...