3,4-Diaminofurazan 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3,4-Diamino-1,2,5-oxadiazole, 1,2,5-Oxadiazole-3,4-diamine

357910010
ACRO357910010EA 48.4 EUR
ACRO357910010
3,4-Diaminofurazan 97%
3,4-Diaminofurazan

Appearance: White to light beige Crystalline powder

Formel: C₂H₄N₄O
Schmelzpunkt: 178…183 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00138084
CAS-Nummer: 17220-38-1

Bestellen

Spezifikation Testergebnisse

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 178 to 183 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR