5,5'-Dimethyl-2,2'-bithiophene 99%

Lieferant: Thermo Scientific

357950010
ACRO357950010EA 69.2 EUR
ACRO357950010
5,5'-Dimethyl-2,2'-bithiophene 99%
5,5'-Dimethyl-2,2'-bithiophene

Appearance: Yellow to light brown Crystals

Formel: C₁₀H₁₀S₂
Schmelzpunkt: 65…68 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01927313
CAS-Nummer: 16303-58-5

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to light brown Crystals
Infrared spectrum Conforms
Melting point 65 to 68 °C
GC ≥99.9 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR