tert-Butyl (4R,6R)-6-cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetate 97%
Lieferant: Thermo Scientific
Synonyme:
(4R,6R)-6-Cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetic Acid tert-Butyl Ester, tert-Butyl 2-((4R,6R)-6-(cyanomethyl)-2,2-dimethyl-1,3-dioxan-4-yl)acetate
357990010
ACRO357990010EA
58.5
EUR
ACRO357990010
tert-Butyl (4R,6R)-6-cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetate 97%
tert-Butyl (4R,6R)-6-cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetate
Appearance: Light yellow Crystalline powder
Formel:
C₁₄H₂₃NO₄ Molecular Weight: 269,34 g/mol Schmelzpunkt: 65…70 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03093958 CAS-Nummer: 125971-94-0 |
Spezifikation Testergebnisse
Appearance | Light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 65 to 70 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...