4-(Aminomethyl)phenylboronsäure Hydrochlorid 96%

Lieferant: Thermo Fisher Scientific
Achtung

358842500
ACRO358842500EA 65.4 EUR
ACRO358842500
4-(Aminomethyl)phenylboronsäure Hydrochlorid 96%
4-(Aminomethyl)phenylboronsäure Hydrochlorid
Formel: C₇H₁₁BClNO₂
Lagertemperatur: Kühlschrank
MDL: MFCD01632199
CAS-Nummer: 75705-21-4

Bestellen

Spezifikation Testergebnisse

Appearance Light yellow Powder
Infrared spectrum Conforms
Titration Argentometric ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR