2-Benzo[b]thiophenboronsäure 98%

Lieferant: Thermo Fisher Scientific
Achtung

358910050
ACRO358910050EA 90.1 EUR
ACRO358910050
2-Benzo[b]thiophenboronsäure 98%
2-Benzo[b]thiophenboronsäure
Formel: C₈H₇BO₂S
Schmelzpunkt: 236…237 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01075674
CAS-Nummer: 98437-23-1

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR