(2-((Dimethylamino)methyl)phenyl)boronic acid 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-(N,N-Dimethylaminomethyl)phenylboronic acid, 2-(Dimethylaminomethyl)phenylboronic acid, 2-(Dimethylaminomethyl)benzeneboronic acid
359241000
ACRO359241000EA
111
EUR
ACRO359241000
(2-((Dimethylamino)methyl)phenyl)boronic acid 98%
(2-((Dimethylamino)methyl)phenyl)boronic acid
CAS No.: 85107-53-5
Formel:
C₉H₁₄BNO₂ Molecular Weight: 179,0240 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD01318999 CAS-Nummer: 85107-53-5 |
Spezifikation Testergebnisse
Appearance | Beige Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...