2-Formylphenylboronsäure 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Boronobenzaldehyd, (2-Formylphenyl)metaborsäure, Dihydroxy(2-formylphenyl)boran, 2-Formylbenzolboronsäure
AC35936.1
AC35936.10
359360010
359360050
MAYBAC35936.1EA
0
EUR
MAYBAC35936.1
MAYBAC35936.10
ACRO359360010
ACRO359360050
2-Formylphenylboronsäure 98%
2-Formylphenylboronsäure
Formel:
C₇H₇BO₃ Molecular Weight: 149.94 g/mol Schmelzpunkt: 120…122 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00151822 CAS-Nummer: 40138-16-7 |
Spezifikation Testergebnisse
Appearance | White to pink to orange Crystalline powder |
Infrared spectrum | Conforms |
Titration with NaOH | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...