3-(Trifluormethyl)phenylboronsäure 97+%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-(Trifluormethyl)benzolboronsäure, Dihydroxy(3-(trifluormethyl)phenyl)boran, (3-(Trifluormethyl)phenyl)metaborsäure
AC35975.1
AC35975.10
359750010
359750050
MAYBAC35975.1EA
0
EUR
MAYBAC35975.1
MAYBAC35975.10
ACRO359750010
ACRO359750050
3-(Trifluormethyl)phenylboronsäure 97+%
3-(Trifluormethyl)phenylboronsäure
Formel:
C₇H₆BF₃O₂ Schmelzpunkt: 160…165 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00151854 CAS-Nummer: 1423-26-3 |
Spezifikation Testergebnisse
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Melting point | 161 to 167 °C |
HPLC | ≥97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...