4-(Trifluormethyl)phenylboronsäure 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (4-(Trifluormethyl)phenyl)metaborsäure, Dihydroxy(4-(trifluormethyl)phenyl)boran, 4-(Trifluormethyl)benzolboronsäure

359765000 359760050
ACRO359765000EA 27.7 EUR
ACRO359765000 ACRO359760050
4-(Trifluormethyl)phenylboronsäure 98%
4-(Trifluormethyl)phenylboronsäure
Formel: C₇H₆BF₃O₂
Molecular Weight: 189.93 g/mol
Schmelzpunkt: 245…250 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00151855
CAS-Nummer: 128796-39-4

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR