4-Thioxo-3,4-dihydro-2(1H)-pyrimidinon 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Hydroxy-4-mercaptopyrimidin
359930010
ACRO359930010EA
98.8
EUR
ACRO359930010
4-Thioxo-3,4-dihydro-2(1H)-pyrimidinon 97%
4-Thiouracil
Formel:
C₄H₄N₂OS Molecular Weight: 128,15 g/mol Schmelzpunkt: 295 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00090842 CAS-Nummer: 591-28-6 |
Spezifikation Testergebnisse
Appearance | Yellow Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Appearance of solution | (5% sol. in 1N NaOH) Clear and faint yellow |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...