Silica gel, functionalized, Amino-3 ∼1,4 mmol/g, Ø 40-63 micron

Lieferant: Thermo Fisher Scientific

360180100 360180250 360181000
ACRO360180100EA 66 EUR
ACRO360180100 ACRO360180250 ACRO360181000
Silica gel, functionalized, Amino-3 ∼1,4 mmol/g, Ø 40-63 micron
Silica gel, functionalized, Amino-3

CAS No.: 71888-97-6

Formel: C₉H₂₃NO₅Si₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03427643
CAS-Nummer: 71888-97-6

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Particle size 40 to 63 µm
Loading ≥1.2 mmol/g (molecular)
Nitrogen (N) ≥1.68 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR