2-N-BOC-amino-3-(4-tetrahydropyranyl)propionic acid 95%

Lieferant: Thermo Fisher Scientific
Achtung

360485000
ACRO360485000EA 478 EUR
ACRO360485000
2-N-BOC-amino-3-(4-tetrahydropyranyl)propionic acid 95%
2-N-BOC-amino-3-(4-tetrahydropyranyl)propionic acid

Appearance: White to off-white Chunks

Formel: C₁₃H₂₃NO₅
Lagertemperatur: Raumtemperatur
MDL: MFCD02683139
CAS-Nummer: 182287-51-0

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Chunks
Infrared spectrum Conforms
NMR ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR