4-(Brommethyl)benzolsulfonylchlorid 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
360540010
360540050
360540250
ACRO360540010EA
25.3
EUR
ACRO360540010
ACRO360540050
ACRO360540250
4-(Brommethyl)benzolsulfonylchlorid 95%
4-(Brommethyl)benzolsulfonylchlorid
Formel:
C₇H₆BrClO₂S Schmelzpunkt: 69…76 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00156129 CAS-Nummer: 66176-39-4 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...