4-(Brommethyl)benzolsulfonylchlorid 95%

Lieferant: Thermo Fisher Scientific

360540010 360540050 360540250
ACRO360540010EA 25.3 EUR
ACRO360540010 ACRO360540050 ACRO360540250
4-(Brommethyl)benzolsulfonylchlorid 95%
4-(Brommethyl)benzolsulfonylchlorid

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 72 to 77 °C
Titration Argentometric ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR