ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Dinatriumsalz, oxidierte Form (NADP-Na2) 97+%

Lieferant: Thermo Fisher Scientific

360565000 360561000
ACRO360565000EA 284 EUR
ACRO360565000 ACRO360561000
ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Dinatriumsalz, oxidierte Form (NADP-Na2) 97+%
ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Dinatriumsalz, oxidierte Form (NADP-Na2)

ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)

Formel: C₂₁H₂₆N₇Na₂O₁₇P₃
Lagertemperatur: Kühlschrank
MDL: MFCD00065390
CAS-Nummer: 24292-60-2
EINECS: 246-129-8

Bestellen

Spezifikation Testergebnisse

Appearance Yellow Powder
Infrared spectrum Conforms
Assay ≥97.0 % (enzymatic, on dry subst.)
Water ≤6.0 %
Solubility (0.05 % in water) Clear colorless to light yellow


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR