3-Bromo-2,5-diaminopyridine 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Bromopyridine-2,5-diamine, 2,5-Diamino-3-bormopyridine, 3-Bromanylpyridine-2,5-diamine
360660010
360660050
ACRO360660010EA
76.3
EUR
ACRO360660010
ACRO360660050
3-Bromo-2,5-diaminopyridine 99%
3-Bromo-2,5-diaminopyridine
Formel:
C₅H₆BrN₃ Schmelzpunkt: 117…122 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03427651 CAS-Nummer: 896160-69-3 |
Spezifikation Testergebnisse
Appearance | Dark brown to violet Powder |
Infrared spectrum | Conforms |
HPLC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...