3-Amino-2-pyridone 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
360710010
360710050
ACRO360710010EA
33.3
EUR
ACRO360710010
ACRO360710050
3-Amino-2-pyridone 97%
3-Amino-2-pyridone
Formel:
C₅H₆N₂O Schmelzpunkt: 118…130 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03411556 CAS-Nummer: 33630-99-8 |
Spezifikation Testergebnisse
Appearance | Dark brown to black Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...