2-Chlor-6-methyl-3-nitropyridin 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 6-Chlor-5-nitro-2-picolin, 6-Chlor-2-methyl-5-nitropyridin, 2-Chlor-3-nitro-6-picolin

360830010 360830050
ACRO360830010EA 60.5 EUR
ACRO360830010 ACRO360830050
2-Chlor-6-methyl-3-nitropyridin 98%
2-Chloro-6-methyl-3-nitropyridine
Formel: C₆H₅ClN₂O₂
Molecular Weight: 172.57 g/mol
Schmelzpunkt: 70…74 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03085820
CAS-Nummer: 56057-19-3
UN: 2811
ADR: 6.1 II

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to brown Powder
Infrared spectrum Conforms
Melting point 69 to 74 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR