4-Aminonicotinsäure 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Aminopyridin-3-carbonsäure, 4-Amino-3-pyridincarbonsäure

361130010 361130050
ACRO361130010EA 27.7 EUR
ACRO361130010 ACRO361130050
4-Aminonicotinsäure 97%
4-Aminonicotinsäure
Formel: C₆H₆N₂O₂
Molecular Weight: 138.13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00234183
CAS-Nummer: 7418-65-7

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to beige Powder
Infrared spectrum Conforms
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR