(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)decanoic acid 95%, 98% ee
Lieferant: Thermo Scientific
Synonyme:
(S)-N-FMOC-Octylglycine, N-Fmoc-2-octyl-L-glycine, Fmoc-octyl-Gly-OH
361490010
ACRO361490010EA
417
EUR
ACRO361490010
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)decanoic acid 95%, 98% ee
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)decanoic acid
Appearance: White Powder
Formel:
C₂₅H₃₁NO₄ Molecular Weight: 409,526 g/mol Schmelzpunkt: 142,6 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01311753 CAS-Nummer: 193885-59-5 |
Spezifikation Testergebnisse
Appearance | White Powder |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Enantiomeric excess | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...