(1S,4R)-N-Boc-1-aminocyclopent-2-ene-4-carboxylic acid 95%, 98% ee

Lieferant: Thermo Scientific
Achtung

Synonyme: (1R,4S)-4-((tert-Butoxycarbonyl)amino)cyclopent-2-enecarboxylic acid

361820010
ACRO361820010EA 164 EUR
ACRO361820010
(1S,4R)-N-Boc-1-aminocyclopent-2-ene-4-carboxylic acid 95%, 98% ee
(1S,4R)-N-Boc-1-aminocyclopent-2-ene-4-carboxylic acid

CAS No.: 151907-80-1

Formel: C₁₁H₁₇NO₄
Molecular Weight: 227,26 g/mol
Schmelzpunkt: 152 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00211288
CAS-Nummer: 151907-80-1

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥94 %
Enantiomeric excess ≥97.5 % (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR