(1S,2R,6S,7R)-4,4-Dimethyl-3,5-dioxa-8-azatricyclo[5.2.1.0(2,6)]decan-9-one 95%, 98% ee

Lieferant: Thermo Scientific
Achtung

362200010
ACRO362200010EA 110 EUR
ACRO362200010
(1S,2R,6S,7R)-4,4-Dimethyl-3,5-dioxa-8-azatricyclo[5.2.1.0(2,6)]decan-9-one 95%, 98% ee
(1S,2R,6S,7R)-4,4-Dimethyl-3,5-dioxa-8-azatricyclo[5.2.1.0(2,6)]decan-9-one

Appearance: White to off-white Crystalline powder or crystals or needles

Formel: C₉H₁₃NO₃
Schmelzpunkt: 150…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01320865
CAS-Nummer: 178032-63-8

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder or crystals or needles
Infrared spectrum Conforms
GC ≥94 %
Enantiomeric excess ≥97.5 % (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR