(1S,2R,3S,4S)-2,3-Dihydroxy-4-(hydroxymethyl)-1-aminocyclopentane hydrochloride 95%, 98% ee

Lieferant: Thermo Scientific
Achtung

Synonyme: (1S,2R,3S,5S)-3-Amino-5-(hydroxymethyl)cyclopentane-1,2-diol hydrochloride, (2S,3S,5S,1R)-5-amino-3-(hydroxymethyl)cyclopentane-1,2-diol, chloride

362530010
ACRO362530010EA 285 EUR
ACRO362530010
(1S,2R,3S,4S)-2,3-Dihydroxy-4-(hydroxymethyl)-1-aminocyclopentane hydrochloride 95%, 98% ee
(1S,2R,3S,4S)-2,3-Dihydroxy-4-(hydroxymethyl)-1-aminocyclopentane hydrochloride
Formel: C₆H₁₄ClNO₃
Molecular Weight: 183,64 g/mol
Schmelzpunkt: 124…124,6 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06799069
CAS-Nummer: 220497-88-1

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
HPLC ≥94 %
Enantiomeric excess ≥97.5 %
Specific optical rotation +25° to +35° (20°C, 589 nm) (c=1 in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR