(R)-N-Boc-allylglycine 95%, 98% ee

Lieferant: Thermo Fisher Scientific
Achtung

362840010 362845000
ACRO362840010EA 158 EUR
ACRO362840010 ACRO362845000
(R)-N-Boc-allylglycine 95%, 98% ee
(R)-N-Boc-allylglycine

Appearance: Clear light yellow Viscous liquid

Formel: C₁₀H₁₆NO₄
Lagertemperatur: Kühlschrank
MDL: MFCD01320882
CAS-Nummer: 170899-08-8

Bestellen

Spezifikation Testergebnisse

Appearance Clear light yellow Viscous liquid or paste
Infrared spectrum Conforms
GC ≥94 %
Enantiomeric excess ≥97.5 %
Specific optical rotation -9.5° to -12.5° (20°C, 589 nm) (c=1, methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR